Thermo Scientific Chemicals

Phthalamide, 97%, Thermo Scientific Chemicals

Catalog number: L08600.18
50 g, Each
Thermo Scientific Chemicals

Phthalamide, 97%, Thermo Scientific Chemicals

Catalog number: L08600.18
50 g, Each
Quantity
Catalog number: L08600.18
also known as L08600-18
Price (USD)
Price: 124.00
Online price: 107.65
Your price:
Quantity
-

Chemical Identifiers

CAS
88-96-0
IUPAC Name
benzene-1,2-dicarboxamide
Molecular Formula
C8H8N2O2
InChI Key
NAYYNDKKHOIIOD-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=CC=C1C(N)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text