Thermo Scientific Chemicals

Dibenzyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: L08813.03
1 g, Each
Thermo Scientific Chemicals

Dibenzyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: L08813.03
1 g, Each
Quantity
Catalog number: L08813.03
also known as L08813-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1623-08-1
IUPAC Name
bis(benzyloxy)phosphinic acid
Molecular Formula
C14H15O4P
InChI Key
HDFFVHSMHLDSLO-UHFFFAOYSA-N
SMILES
OP(=O)(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
76.0-83.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text