Thermo Scientific Chemicals

4-Chlorobenzyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L09579.06
5 g, Each
Thermo Scientific Chemicals

4-Chlorobenzyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: L09579.06
5 g, Each
Quantity
Catalog number: L09579.06
also known as L09579-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
3694-45-9
IUPAC Name
1-chloro-4-(isothiocyanatomethyl)benzene
Molecular Formula
C8H6ClNS
InChI Key
DEHXIHUIYSXZNH-UHFFFAOYSA-N
SMILES
ClC1=CC=C(CN=C=S)C=C1
Assay (GC)
≥96.0%
Refractive Index
1.6120-1.6170 @ 20°C
Form
Liquid
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text