Thermo Scientific Chemicals

4-Hydroxybutyl acrylate, 95%, stab. with ca 500ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L11097.14
25 g, Each
Thermo Scientific Chemicals

4-Hydroxybutyl acrylate, 95%, stab. with ca 500ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L11097.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L11097.14
also known as L11097-14
Price (USD)
Price: 135.00
Your price:
Quantity
-

Chemical Identifiers

CAS
2478-10-6
IUPAC Name
4-hydroxybutyl prop-2-enoate
Molecular Formula
C7H12O3
InChI Key
NDWUBGAGUCISDV-UHFFFAOYSA-N
SMILES
OCCCCOC(=O)C=C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%
Form
Liquid
Refractive Index
1.4510-1.4550 @ 20?C

Description

4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybutyl acrylate is used in paint and coating materials (excellent scratch resistance, mechanical properties, and acid rain resistance ), pressure sensitive adhesives, UV/EB curable composition, waterborne coatings.

Solubility
Fully miscible in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text