Thermo Scientific Chemicals

Bis(2-hydroxyethyl) disulfide, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11800.30
250 g, Each
Thermo Scientific Chemicals

Bis(2-hydroxyethyl) disulfide, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11800.30
250 g, Each
Quantity
Catalog number: L11800.30
also known as L11800-30
Price (USD)
Price: 218.00
Online price: 185.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1892-29-1
IUPAC Name
2-[(2-hydroxyethyl)disulfanyl]ethan-1-ol
Molecular Formula
C4H10O2S2
InChI Key
KYNFOMQIXZUKRK-UHFFFAOYSA-N
SMILES
OCCSSCCO
Assay (GC)
≥88.0%
Form
Liquid
Water Content (Karl Fischer Titration)
≤10.0%
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms

Description

Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.

Solubility
Soluble in water, ethanol, acetone, and ether.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text