3,5-Di-tert-butylbenzoic acid is used in the synthesis of 5,5-di-tert-butylanilinium sulfate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
3,5-Di-tert-butylbenzoesäure wird bei der Synthese von 5,5-Di-tert-butyliniumsulfat verwendet.
Löslichkeit
Unlöslich in Wasser.
Hinweise
Nicht kompatibel mit Oxidationsmitteln. In einem gut abgedichteten Behälter kühl und trocken lagern.
RUO – Research Use Only
John W. Elder; Raymond P. Mariella. The Synthesis and Substitution Reactions of 3,5-Di-Tert-Butylphenol. Canadian Journal of Chemistry.2002, 41, (7), 1653-1656.