4-(Trifluoromethyl)phenyl isocyanate has been used in the synthesis of fluorescent photoinduced electron transfer (PET) chemosensors and 2-ethyl-6-[(4-trifluoromethylphenylcarbamoyl)hydrazino]-benzo[de]iso-quinoline-1,3-dione, an amidourea-based sensor for anions. It has also been used in the synthesis of 1,8-naphthalimide based colorimetric and fluorescent sensor for anions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
4-(Trifluormethyl)phenylisocyanat wird bei der Synthese von Chemosensoren für den fluoreszierenden photoinduzierten Elektronentransfer (PET) und von 2-Ethyl-6[(4-trifluormethylphenylcarbamoyl)hydrazino]-benzo[de]iso-chinolin-1,3-dion, einem auf Imidurea basierenden Sensor für Anionen, verwendet. Es wird auch bei der Synthese von 1,8 kolorimetrischen und fluoreszierenden auf Naphthalimid basierenden Anionensensoren verwendet.
Löslichkeit
Es hydrolysiert in Wasser.
Hinweise
Feuchtigkeitsempfindlich. Nicht kompatibel mit Oxidationsmitteln, Alkoholen, Aminen und Hitze.
RUO – Research Use Only
Allgemeine Referenzen:
- Thorfinnur Gunnlaugsson; Anthony P. Davis; John E. O'Brien; Mark Glynn. Synthesis and photophysical evaluation of charge neutral thiourea or urea based fluorescent PET sensors for bis-carboxylates and pyrophosphate. Organic & Biomolecular Chemistry. 2005, 3,(1), 48-56.
- Thorfinnur Gunnlaugsson; Paul E. Kruger; Paul Jensen; Juliann Tierney; Haslin Dato Paduka Ali; Gillian M. Hussey. Colorimetric naked eye sensing of anions in aqueous solution. J. Org. Chem. 2005, 70,(26), 10875-10878.