Thermo Scientific Chemicals

4-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L13120.22
100 g, Each
Thermo Scientific Chemicals

4-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L13120.22
100 g, Each
Quantity
Catalog number: L13120.22
also known as L13120-22
Price (USD)
Price: 117.00
Online price: 99.65
Your price:
Quantity
-

Chemical Identifiers

CAS
100-07-2
IUPAC Name
4-methoxybenzoyl chloride
Molecular Formula
C8H7ClO2
InChI Key
MXMOTZIXVICDSD-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(Cl)=O
Assay (GC)
≥96.0%
Appearance (Color)
Colorless to white to yellow
Identification (FTIR)
Conforms
Form
Fused solid or clear liquid as melt
Refractive Index
1.5790-1.5840 @ 20?C

Description

4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.

Solubility
It reacts in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text