Thermo Scientific Chemicals

Hexanes, mixed isomers, 98+%, Thermo Scientific Chemicals

Catalog number: L13233.0F
2500 mL, Each
Thermo Scientific Chemicals

Hexanes, mixed isomers, 98+%, Thermo Scientific Chemicals

Catalog number: L13233.0F
2500 mL, Each
Quantity
Catalog number: L13233.0F
also known as L13233-0F
Price (USD)
Price: 74.00
Online price: 62.65
Your price:
Quantity
-

Chemical Identifiers

CAS
92112-69-1
IUPAC Name
hexane
Molecular Formula
C6H14
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
Refractive Index
1.3775-1.3815 @ 20?C
Assay (GC)
≥98.0% (sum of isomers)
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

Hexane mixed isomers are used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds, namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexane mixed isomers are used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds, namely butyl lithium.

Solubility
Miscible with alcohol, chloroform, acetone and ether. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents. Keep away from sources of ignition.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text