Thermo Scientific Chemicals

Dowex™ 50WX4 200-400 (H), Thermo Scientific Chemicals

Catalog number: L13918.30
250 g, Each
Thermo Scientific Chemicals

Dowex™ 50WX4 200-400 (H), Thermo Scientific Chemicals

Catalog number: L13918.30
250 g, Each
Quantity
Catalog number: L13918.30
also known as L13918-30
Price (USD)
Price: 158.00
Your price:
Quantity
-

Chemical Identifiers

CAS
11113-61-4
MDL Number
MFCD00132725
Appearance (Color)
White to pale brown
Exchange capacity
>1.1 eq/L (H)
Moisture holding capacity
64-72% (H)
Form
Beads
Matrix
Sulfonic acid active functional group

Description

Dowex™ 50WX4 200-400 (H) is used to investigate the effect of particle size on drug loading and release profiles. Dowex™ 50WX4 hydrogen form when treated in boiling water converted D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) to free aldehyde D,L-glyceraldehyde 3-phosphate. It is a useful column packing material, used in anion exchange chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex™ 50WX4 200-400 (H) is used to investigate the effect of particle size on drug loading and release profiles. Dowex™ 50WX4 hydrogen form when treated in boiling water converted D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) to free aldehyde D,L-glyceraldehyde 3-phosphate. It is a useful column packing material, used in anion exchange chromatography.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text