Thermo Scientific Chemicals

Pyromellitic diimide, 97%, Thermo Scientific Chemicals

Catalog number: L14372.14
25 g, Each
Thermo Scientific Chemicals

Pyromellitic diimide, 97%, Thermo Scientific Chemicals

Catalog number: L14372.14
25 g, Each
Quantity
Catalog number: L14372.14
also known as L14372-14
Price (USD)
Price: 148.00
Online price: 125.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2550-73-4
IUPAC Name
1H,2H,3H,5H,6H,7H-pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
Molecular Formula
C10H4N2O4
InChI Key
UGQZLDXDWSPAOM-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C2=CC3=C(C=C12)C(=O)NC3=O
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White or cream to pale brown or pale pink to pale purple
Assay (HPLC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text