Thermo Scientific Chemicals

L-Proline benzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L15618.03
1g, Each
Thermo Scientific Chemicals

L-Proline benzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L15618.03
1g, Each
Quantity
Catalog number: L15618.03
also known as L15618-03
Price (USD)

Chemical Identifiers

CAS
16652-71-4
IUPAC Name
benzyl pyrrolidine-2-carboxylate hydrochloridyl
Molecular Formula
C12H15ClNO2
InChI Key
MRWGIRHDSFQISP-UHFFFAOYNA-N
SMILES
[Cl].O=C(OCC1=CC=CC=C1)C1CCCN1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5% (UK Sourced Material)
Assay from Supplier's CofA
≥97.5% (HPLC, US Sourced Material)
Comment
Material Sourced in UK and US

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text