Thermo Scientific Chemicals

Dimethyl sebacate, 97%, Thermo Scientific Chemicals

Catalog number: L15805.36
500 g, Each
Thermo Scientific Chemicals

Dimethyl sebacate, 97%, Thermo Scientific Chemicals

Catalog number: L15805.36
500 g, Each
Quantity
Catalog number: L15805.36
also known as L15805-36
Price (USD)
Price: 179.00
Online price: 154.65
Your price:
Quantity
-

Chemical Identifiers

CAS
106-79-6
IUPAC Name
1,10-dimethyl decanedioate
Molecular Formula
C12H22O4
InChI Key
ALOUNLDAKADEEB-UHFFFAOYSA-N
SMILES
COC(=O)CCCCCCCCC(=O)OC
Refractive Index
1.4360-1.4410 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Form
Liquid

Description

Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text