Thermo Scientific Chemicals

Allyl isocyanate, 96%, Thermo Scientific Chemicals

Catalog number: L18710.03
1 g, Each
Thermo Scientific Chemicals

Allyl isocyanate, 96%, Thermo Scientific Chemicals

Catalog number: L18710.03
1 g, Each
Quantity
Catalog number: L18710.03
also known as L18710-03
Price (USD)
Price: 35.80
Online price: 30.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1476-23-9
Assay from Supplier's CofA
≥95.0%
Infrared spectrum
Conforms
Appearance (Color)
Clear or slightly turbid, colorless to yellow
Comment
May form small amount of sediment upon storage.
Form
Liquid

Description

It finds its application in the synthesis, characterization of biodegradable dextran-allyl isocyanate-ethylamine/polyethylene glycol-diacrylate hydrogels and their in vitro release of albumin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis, characterization of biodegradable dextran-allyl isocyanate-ethylamine/polyethylene glycol-diacrylate hydrogels and their in vitro release of albumin.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text