Thermo Scientific Chemicals

Spermine, 97%, Thermo Scientific Chemicals

Catalog number: L19562.06
5 g, Each
Thermo Scientific Chemicals

Spermine, 97%, Thermo Scientific Chemicals

Catalog number: L19562.06
5 g, Each
Quantity
Catalog number: L19562.06
also known as L19562-06
Price (USD)
Price: 188.00
Online price: 159.65
Your price:
Quantity
-

Chemical Identifiers

CAS
71-44-3
IUPAC Name
(3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine
Molecular Formula
C10H26N4
InChI Key
PFNFFQXMRSDOHW-UHFFFAOYSA-N
SMILES
NCCCNCCCCNCCCN
Appearance (Color)
White
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Water Content (Karl Fischer Titration)
≤1.0%
Identification (FTIR)
Conforms (non-U.S. specification)

Description

Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Binds to the polyamine modulatory site of NMDA Spermine is essential for both normal and neoplastic tissue growth. It is involved in the modulation of calcium-dependent immune processes. It plays an important role in cellular proliferation and differentiation as well as inhibits neuronal nitric oxide synthase (nNOS).

Solubility
Soluble in chloroform, methanol, water and lower alcohols. Insoluble in ether, benzene and petroleum ether.

Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text