Thermo Scientific Chemicals

Methyl 3,3-dimethylacrylate, 98%, Thermo Scientific Chemicals

Catalog number: L19731.09
10 g, Each
Thermo Scientific Chemicals

Methyl 3,3-dimethylacrylate, 98%, Thermo Scientific Chemicals

Catalog number: L19731.09
10 g, Each
Quantity
Catalog number: L19731.09
also known as L19731-09
Price (USD)
Price: 73.20
Online price: 62.65
Your price:
Quantity
-

Chemical Identifiers

CAS
924-50-5
IUPAC Name
methyl 3-methylbut-2-enoate
Molecular Formula
C6H10O2
InChI Key
FZIBCCGGICGWBP-UHFFFAOYSA-N
SMILES
COC(=O)C=C(C)C
Form
Liquid
Refractive Index
1.4360-1.4400 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in chloroform. Insoluble in water,

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is sensitive to light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text