Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™
Avantor™

Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
2.5 kg
12 kg
110 lb.
220 lb.
10 x 10 kg
Verpackung:
Alle anzeigen
Polyethylene pail, screw top
Katalognummer M7713-30
Preis (EUR)
-
Menge:
10 x 10 kg
Verpackung:
Polyethylene pail, screw top
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Specifications
Siedepunkt1413°C (1013 hPa)
CAS7647-14-5
Chemischer Name oder MaterialSodium chloride
FarbeWeiß
Schmelzpunkt801°C
SummenformelNaCl
VerpackungPolyethylene pail, screw top
SynonymSalt, Halite, Sea salt, Rock salt, Saline
GüteUSP
Molekulargewicht (g/mol)58.44
Reinheit (%)99.0-100.5%
Menge10 x 10 kg
Unit SizeEach