Oregon Green&trade; 488 1,2-Dihexadecanoyl-<i>sn</i>-Glycero-3-Phosphoethanolamine (Oregon Green&trade; 488 DHPE)
Invitrogen™

Oregon Green™ 488 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Oregon Green™ 488 DHPE)

リン脂質であるOregon Green™ 488 DHPEは、励起/最大発光約501/526 nmを持つ緑色蛍光Oregon Green™フルオロフォアで、ヘッドグループ上に標識されています。このフッ素化されたフルオレセインのアナログは、光安定性の向上やpKaの低下詳細を見る
製品番号(カタログ番号)数量
O12650
または、製品番号O-12650
1 mg
製品番号(カタログ番号) O12650
または、製品番号O-12650
価格(JPY)
33,400
キャンペーン価格
Ends: 25-Sep-2026
55,700
割引額 22,300 (40%)
Each
数量:
1 mg
リン脂質であるOregon Green™ 488 DHPEは、励起/最大発光約501/526 nmを持つ緑色蛍光Oregon Green™フルオロフォアで、ヘッドグループ上に標識されています。このフッ素化されたフルオレセインのアナログは、光安定性の向上やpKaの低下(フルオレセインではpKa∼ 4.7対6.4)など、フルオレセインのキーとなる限界を克服し、蛍光は生理学的pH範囲で基本的に影響を受けません。
研究用にのみ使用できます。診断用には使用いただけません。
仕様
化学物質名または材質Phospholipids
推奨保存方法Store in freezer (-5 to -30°C) and protect from light.
物理的フォームSolid
製品ラインOregon Green
数量1 mg
Unit SizeEach

引用および参考文献 (7)

引用および参考文献
Abstract
Zero mode waveguides for single-molecule spectroscopy on lipid membranes.
Authors:Samiee KT, Moran-Mirabal JM, Cheung YK, Craighead HG
Journal:Biophys J
PubMed ID:16461393
'Zero mode waveguides (ZMWs), subwavelength optical nanostructures with dimensions ranging from 50 to 200 nm, have been used to study systems involving ligand-receptor interactions. We show that under proper conditions, lipid membranes will invaginate into the nanostructures, which confine optical excitation to subattoliter volumes. Fluorescence correlation spectroscopy (FCS) was used ... More
Early endosomal SNAREs form a structurally conserved SNARE complex and fuse liposomes with multiple topologies.
Authors:Zwilling D, Cypionka A, Pohl WH, Fasshauer D, Walla PJ, Wahl MC, Jahn R
Journal:EMBO J
PubMed ID:17159904
'SNARE proteins mediate membrane fusion in eukaryotic cells. They contain conserved SNARE motifs that are usually located adjacent to a C-terminal transmembrane domain. SNARE motifs spontaneously assemble into four helix bundles, with each helix belonging to a different subfamily. Liposomes containing SNAREs spontaneously fuse with each other, but it is ... More
Surface-coupled proton exchange of a membrane-bound proton acceptor.
Authors:Sandén T, Salomonsson L, Brzezinski P, Widengren J,
Journal:Proc Natl Acad Sci U S A
PubMed ID:20160117
Proton-transfer reactions across and at the surface of biological membranes are central for maintaining the transmembrane proton electrochemical gradients involved in cellular energy conversion. In this study, fluorescence correlation spectroscopy was used to measure the local protonation and deprotonation rates of single pH-sensitive fluorophores conjugated to liposome membranes, and the ... More
Effects of linker sequences on vesicle fusion mediated by lipid-anchored DNA oligonucleotides.
Authors:Chan YH, van Lengerich B, Boxer SG,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19164559
Synthetic lipid-oligonucleotide conjugates inserted into lipid vesicles mediate fusion when one population of vesicles displays the 5'-coupled conjugate and the other the 3'-coupled conjugate, so that anti-parallel hybridization allows the membrane surfaces to come into close proximity. Improved assays show that lipid mixing proceeds more quickly and to a much ... More
Interplay between H1N1 influenza a virus infection, extracellular and intracellular respiratory tract pH, and host responses in a mouse model.
Authors:
Journal:PLoS One
PubMed ID:33979408