Propidium Iodide - FluoroPure™ Grade
Invitrogen™

Propidium Iodide - FluoroPure™ Grade

Because it is manufactured at our ISO 13485—certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade propidium iodideRead more
Have Questions?
Catalog NumberQuantity
P21493100 mg
Catalog number P21493
Price (USD)
275.00
Each
Quantity:
100 mg
Price (USD)
275.00
Each
Because it is manufactured at our ISO 13485—certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade propidium iodide is greater than or equal to 98% pure by HPLC.

Also available as a room-temperature-stable, ready-to-use solution: Propidium Iodide Ready Probes Reagent.
See other Ready Probes ready-to-use imaging reagents and accessories ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPropidium Iodide - FluoroPure™ Grade
Detection MethodFluorescence
Dye TypeCell-Impermeant
Emission Wavelength617
Excitation Wavelength535 nm
Quantity100 mg
Shipping ConditionRoom Temperature
Sub Cellular LocalizationCytoplasm and Cytosol
ColorRed
For Use With (Application)Viability Assay
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineFluoroPure
Product TypePropidium Iodide
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.

Frequently asked questions (FAQs)

I have a LIVE/DEAD BacLight Bacterial Viability kit that has SYTO 9 and propidium iodide in it. Will I be able to stain eukaryotic cells that have engulfed bacteria and determine if the bacteria are alive or dead using this kit?

Unfortunately, no. SYTO 9 will label the nuclei of live or dead cells, including the eukaryotic cells. Propidium iodide is cell impermeant, and will only enter dead cells. If the eukaryotic cells are dead, they will label with propidium iodide as well. If the eukaryotic cells are alive, propidium iodide will not be able to enter and thus will not label the bacteria inside, whether the bacteria are alive or dead. We are not aware of any way to do a viability assay of bacteria once they have been engulfed by cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is propidium iodide (PI) fixable with glutaraldehyde or paraformaldehyde (PFA)?

PI is not fixable with glutaraldehyde or PFA. Both reagents fix by crosslinking amines. PI and other nucleic acid stains do not inherently bind covalently to nucleic acids and these fixatives do not crosslink the dyes to nucleic acids.
The one fixable nucleic acid stain is Ethidium Monoazide Bromide (EMA), Cat no. E1374); it covalently binds to nucleic acids upon activation by exposure to light.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What kinds of cell health and viability assays can be performed by flow cytometry?

The following cell health and viability assays can be performed by flow cytometry :

-Apoptosis Assays:
Membrane Asymmetry: Annexin V is a member of a family of structurally related proteins that bind phospholipids in the presence of Ca2+. Annexin V binds several phospholipids, but shows highest affinity for phosphatidylserine.
Phosphatidylserine is normally found in the inner leaflet of the cell membrane; however, in the early stages of apoptosis, phosphatidylserine is observed to translocate to the outer leaflet. This translocation makes phosphatidylserine available for annexin V binding in the presence of Ca2+ containing incubation buffer. Cells undergoing apoptosis will stain with annexin V, while normal cells will not. annexin V is available conjugated with a wide range of fluorophores.

Mitochondrial Health: A distinctive feature of the early stages of apoptosis is the disruption of the mitochondria, including changes in membrane and redox potential. We exclusively offer a number of fluorescent probes for analyzing mitochondrial activity in live cells by flow cytometry, with minimal disruption of cellular function.

The MitoProbe family of mitochondrial stains (MitoProbe DiOC2(3) Assay Kit, Cat. No. M34150, MitoProbe JC-1 Assay Kit, Cat. No. M34152, and MitoProbe DiIC1(5) Assay Kit, Cat. No. M34151) provides quick, easy, and reliable flow cytometric detection of the loss of mitochondrial membrane potential that occurs during apoptosis.

Caspase Activity: The CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit (Cat. No. C10427) enables flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic cells. The kit includes the novel fluorogenic substrate CellEvent Caspase-3/7 Green Detection Reagent which targets the recognition sequence for activated caspase-3 and caspase-7, as well as SYTOX AADvanced Dead Cell Stain.

DNA Fragmentation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. DNA fragmentation that occurs during apoptosis produces DNA strand breaks, and can be analyzed using TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) assays. The APO-BrdU TUNEL assay (Cat. No. A23210) is a two-color assay for labeling DNA breaks and total cellular DNA to detect apoptotic cells by imaging or flow cytometry.

Nuclear Chromatin Condensation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. Cells undergoing apoptosis display an increase in nuclear chromatin condensation. As the chromatin condenses, cell-permeable nucleic acid stains becomes hyperfluorescent, thus enabling the identification of apoptotic cells when combined with a traditional dead-cell stain. The Vybrant Apoptosis Assay Kit #5, Hoechst 33342/Propidium Iodide (Cat. No. V13244) provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. The Chromatin Condensation & Membrane Permeability Dead Cell Apoptosis Kit with Hoechst 33342, YO-PRO-1, and PI dyes, for flow cytometry (Cat. No. V23201) detects apoptotic cells with changes in nuclear chromatin condensation and plasma membrane permeability.

-Cell Cycle Analysis:
Live cell assays: The Vybrant DyeCycle family of dyes offers robust fluorescent dyes for live-cell cycle analysis with limited cytotoxicity using 405 nm (Cat. No. V35003), 488 nm (Cat. No. V35004), 532 nm (Cat. No. V35005), or 633 nm (Cat. Nos. V10309 and V10273) excitation. The dyes have low cytotoxicity, allowing stained cells to be sorted and otherwise cultured or assessed with functional assays after staining.

Fixed cell assays: Analyzing cell cycle using FxCycle Violet Stain (Cat. No. F10347), SYTOX AADvanced Dead Cell Stain Kit (Cat. No. S10349) or FxCycle Far Red Stain (Cat. No. F10348) allows for multiple color options for simplified fixed cell cycle analysis.

-Cell Proliferation:
Dye dilution assays for cell proliferation: Dye dilution assays for cell proliferation rely on cell membrane–permeant fluorescent molecules. Upon entry into the cell, the dye will covalently bind to amine groups on proteins, resulting in long-term dye retention within the cell. Through subsequent cell divisions, each daughter cell receives approximately half the fluorescence of the parent. Analysis of the fluorescence intensities of cell populations by flow cytometry enables determination of the number of generations through which a cell or population has progressed since the label was applied. CellTrace fluorescent stains can be used without affecting morphology or physiology to trace generations in vivo or in vitro. There is no known effect on proliferative ability or biology of cells and they are well retained in cells for several days post-stain. Available kits for flow cytometry include CellTrace CFSE Cell Proliferation Kit (Cat. No. C34554), CellTrace Violet Cell Proliferation Kit (Cat. No. C34557), and CellTrace Far Red Cell Proliferation Kit (Cat. No. C34564).

DNA Synthesis Assays: Measuring the synthesis of new DNA is a precise way to assay cell proliferation in individual cells or in cell populations. DNA synthesis–based cell proliferation assays measure the rate of new DNA synthesis based on incorporation of modified nucleosides. The Click-iT Plus EdU cell proliferation assay utilizes the power of click chemistry and the modified nucleoside EdU to provide a superior alternative to BrdU staining for detecting and quantitating newly synthesized DNA. The Click-iT Plus EdU cell proliferation assay is available with Pacific Blue (Cat. No. C10636), Alexa Fluor 488 (Cat. Nos. C10632 and C10633), and Alexa Fluor 647 (Cat. Nos. C10634 and C10635).

-Viability Assays:
Dead cells often give false positive results, as they tend to bind non-specifically to many reagents. Therefore, removing dead cells from your flow cytometry data is a critical step to help ensure accurate results and analysis.

Non-fixable Membrane Permeability Stains: SYTOX Dead Cell Stains (Cat. Nos. S34857, S34860, S34861, S34859, and S34862) do not cross intact cell membranes, and they exhibit increased fluorescence upon dsDNA binding, making them some of our most brilliant dead cell stains. Cell-impermeant classic DNA-binding dyes include propidium iodide (Cat. No. P21493) and 7-AAD (Cat. No. A1310). Both of these dyes have been used extensively for viability assays in flow cytometry. CellTrace Calcein AM dyes can be passively loaded into adherent and nonadherent cells. These cell-permeant esterase substrates serve as viability probes that measure both enzymatic activity, which is required to activate their fluorescence, and cell membrane integrity, which is required for intracellular retention of their fluorescent products. Available with blue (Cat. No. C34853), violet (Cat. No. C34858), and green (Cat. No. C34852) fluorescence, these dyes are ideal for short-term staining of live cells and can be used in multiplexed flow cytometry experiments.

Fixable Viability Stains: The LIVE/DEAD Fixable Dead Cell Stains are fixable viability dyes that help to ensure accurate assessment of cell viability in samples after fixation and/or permeabilization. LIVE/DEAD Fixable Dead Cell Stain Kits are based on the reaction of a fluorescent reactive dye with cellular proteins (amines). These dyes cannot penetrate live-cell membranes, so only cell-surface proteins are available to react with the dye, resulting in dim staining. The reactive dye can permeate the damaged membranes of dead cells and stain both the interior and exterior amines, resulting in more intense staining. LIVE/DEAD Fixable Dead Cell Stain Kits are available in eight single-channel colors available for UV, 405, 488, 532, 561, or 633 nm lasers in three packaging sizes to match your experiment.



Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (1094)

Citations & References
Abstract
Signaling through MHC class II molecules blocks CD95-induced apoptosis.
Authors:Catlett IM,Xie P,Hostager BS,Bishop GA
Journal:Journal of immunology (Baltimore, Md. : 1950)
PubMed ID:11342618
Glutamate and non-glutamate receptor mediated toxicity caused by oxygen and glucose deprivation in organotypic hippocampal cultures.
Authors:Newell DW, Barth A, Papermaster V, Malouf AT
Journal:J Neurosci
PubMed ID:7472521
In vitro ischemia models have utilized oxygen, or oxygen and glucose deprivation to simulate ischemic neuronal injury. Combined oxygen and glucose deprivation can induce neuronal damage which is in part mediated through NMDA receptors. Severe oxygen deprivation alone however can cause neuronal injury which is not NMDA mediated. We tested ... More
Identification and characterization of two subpopulations of Encephalitozoon intestinalis.
Authors:Hoffman RM, Marshall MM, Polchert DM, Jost BH
Journal:Appl Environ Microbiol
PubMed ID:12902292
Microsporidia are obligate intracellular protozoa that have been shown to be pathogenic to most living creatures. The development of in vitro cell culture propagation methods has provided researchers with large numbers of spores and facilitated the study of these organisms. Here, we describe heterogeneity within cell culture-propagated Encephalitozoon intestinalis suspensions. ... More
Autoantigens targeted in systemic lupus erythematosus are clustered in two populations of surface structures on apoptotic keratinocytes.
Authors:Casciola-Rosen LA, Anhalt G, Rosen A
Journal:J Exp Med
PubMed ID:7511686
'Systemic lupus erythematosus is a multisystem autoimmune disease in which the autoantibody response targets a variety of autoantigens of diverse subcellular location. We show here that these autoantigens are clustered in two distinct populations of blebs at the surface of apoptotic cells. The population of smaller blebs contains fragmented endoplasmic ... More
Caspase activation contributes to delayed death of heat-stressed striatal neurons.
Authors:White MG, Emery M, Nonner D, Barrett JN
Journal:J Neurochem
PubMed ID:14622126
'Hyperthermia can contribute to brain damage both during development and post-natally. We used rat embryonic striatal neurons in culture to study mechanisms underlying hyperthermia-induced neuronal death. Heat stress at 43 degrees C for 2 h produced no obvious signs of damage during the first 12 h after the stress, but ... More