Propidium Iodide ReadyProbes™ Reagent
Invitrogen™

Propidium Iodide ReadyProbes™ Reagent

Propidium iodide is a commonly used dead-cell stain that emits red fluorescence when bound to DNA. Propidium iodide is alsoRead more
Have Questions?
Catalog NumberQuantity
R371081 kit
Catalog number R37108
Price (USD)
306.68
Special offer
Online exclusive
Ends: 31-Dec-2026
374.00
Save 67.32
Each
Quantity:
1 kit
Price (USD)
306.68
Special offer
Online exclusive
Ends: 31-Dec-2026
374.00
Save 67.32
Each
Propidium iodide is a commonly used dead-cell stain that emits red fluorescence when bound to DNA. Propidium iodide is also used as a counterstain in multicolor fluorescent techniques. With Propidium Iodide ReadyProbes® Reagent, we provide a room temperature-stable solution of classic DNA stain that comes in a convenient-to-use dropper bottle. Just tip and drip two drops per ml to stain your cells.

Also available: Propidium Iodide, Fluorspar grade at 12.5X concentration (13.2 μg/mL, 19.7 μM) in water with 1% DMSO.

See other Ready Probes ready-to-use imaging reagents and accessories ›

Features of Propidium Iodide ReadyProbes® Reagent:
• Ready-to-use liquid propidium iodide formulation
• Rapid staining of dead cells without wash steps
• Ready-to-use liquid formulation in convenient dropper bottle—no need to dilute, weigh, or pipette
• Stable at room temperature—keep handy at your work station or cell culture area

Classic Dead Cell Stain
Propidium iodide is a classic fluorescent dye used extensively for staining of dead cells (Figure 1). It exhibits a sufficiently large Stokes shift to allow simultaneous detection of nuclear DNA and FITC or Alexa Fluor® 488-labeled antibodies.

Packaged for Convenience
Propidium Iodide ReadyProbes® Reagent is supplied ready-to-use and can be stored right next to your flow cytometer or in your cell culture lab. A liquid formulation in an ultra-convenient dropper bottle (Figure 2) allows you to stain your cells whenever and wherever they are ready. Just add two drops per ml for optimally bright and distinct nuclear counter-staining.

Multiple Applications
Propidium iodide is membrane-impermeant and excluded from viable cells. It is therefore commonly used for identifying dead cells in a population. Its spectral properties, selectivity for dsDNA, and increased fluorescence when bound to DNA, make propidium iodide ideal for use with green (Alexa Fluor® 488, FITC, GFP) fluorophores in multicolor experiments. Because of its high affinity for DNA, it is also frequently used for counting cells, measuring apoptosis, and sorting cells based on DNA content.

Suggestions for Use
• In most cases, 2 drops/1×106 cells of Propidium Iodide ReadyProbes® Reagent and an incubation time of 15 to 30 minutes will produce bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
• When bound to nucleic acids, the fluorescence excitation maximum for propidium iodide is 535 nm and the emission maximum is 617 nm. Propidium iodide can be excited with a xenon or mercury-arc lamp or with the 488 line of an argon-ion laser. Generally, propidium iodide fluorescence is detected in the FL2 channel of flow cytometers.
• As a preferred red nuclear stain in dead cell analyses using flow cytometry, Propidium Iodide ReadyProbes® Reagent is ideal for use with antibody-based applications.

See other Molecular Probes® imaging tools and reagents.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-impermeant
Detection MethodFluorescence
Dye Typepropidium iodide
Emission Wavelength617
Excitation Wavelength535 nm
FormLiquid
Quantity1 kit
Sub Cellular LocalizationNucleus
ColorFar-red
For Use With (Application)Viability Assay
For Use With (Equipment)Attune Acoustic Focusing Cytometer, EVOS Cell Imaging System, Flow Cytometer
Product LineReadyProbes
Product TypeNucleic Acid Stain
Unit SizeEach
Contents & Storage
6 x 2.5 mL dropper bottles

Store at ≤ 25°C

Frequently asked questions (FAQs)

What is the volume for one drop from the ReadyProbes dropper bottles?

The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (15)

Citations & References
Abstract
Extension of Saccharomyces paradoxus chronological lifespan by retrotransposons in certain media conditions is associated with changes in reactive oxygen species.
Authors:VanHoute D, Maxwell PH,
Journal:
PubMed ID:25106655
'Retrotransposons are mobile DNA elements present throughout eukaryotic genomes that can cause mutations and genome rearrangements when they replicate through reverse transcription. Increased expression and/or mobility of retrotransposons has been correlated with aging in yeast, Caenorhabditis elegans, Drosophila melanogaster, and mammals. The many copies of retrotransposons in humans and various ... More
Mutation in VPS35 associated with Parkinson's disease impairs WASH complex association and inhibits autophagy.
Authors:Zavodszky E, Seaman MN, Moreau K, Jimenez-Sanchez M, Breusegem SY, Harbour ME, Rubinsztein DC,
Journal:
PubMed ID:24819384
'Endosomal protein sorting controls the localization of many physiologically important proteins and is linked to several neurodegenerative diseases. VPS35 is a component of the retromer complex, which mediates endosome-to-Golgi retrieval of membrane proteins such as the cation-independent mannose 6-phosphate receptor. Furthermore, retromer is also required for the endosomal recruitment of ... More
Modulation of medium pH by caulobacter crescentus facilitates recovery from uranium-induced growth arrest.
Authors:Park DM, Jiao Y,
Journal:
PubMed ID:25002429
The oxidized form of uranium [U(VI)] predominates in oxic environments and poses a major threat to ecosystems. Due to its ability to mineralize U(VI), the oligotroph Caulobacter crescentus is an attractive candidate for U(VI) bioremediation. However, the physiological basis for U(VI) tolerance is unclear. Here we demonstrated that U(VI) caused ... More
Role of betulinic acid derivative SH-479 in triple negative breast cancer and bone microenvironment.
Authors:
Journal:Oncol Lett
PubMed ID:34188707
Sacrificial-layer free transfer of mammalian cells using near infrared femtosecond laser pulses.
Authors:Zhang J, Hartmann B, Siegel J, Marchi G, Clausen-Schaumann H, Sudhop S, Huber HP,
Journal:PLoS One
PubMed ID:29718923
'Laser-induced cell transfer has been developed in recent years for the flexible and gentle printing of cells. Because of the high transfer rates and the superior cell survival rates, this technique has great potential for tissue engineering applications. However, the fact that material from an inorganic sacrificial layer, which is ... More