Edwards Medium with Sheep Blood
Thermo Scientific™

Edwards Medium with Sheep Blood

Unlock the power of precision in microbial cultivation with PB5080A Edwards Medium with Sheep Blood.
Have Questions?
KatalognummerMenge
PB5080A10 Platten
Katalognummer PB5080A
Preis (EUR)
-
Menge:
10 Platten
This high-performance agar medium is specifically designed to support the growth and isolation of a wide range of microorganisms, ensuring accurate and reliable results for your microbiological testing needs.
Specifications
FarbeRed
Füllvolumen17 mL
Format90 mm
Verpackungsart1 x 10 Plates
Menge10 Platten
Haltbarkeit56 Weeks
HerstellungsortUnited Kingdom
ProdukttypAgar Plate
Unit SizeEach
Inhalt und Lagerung
2°C to 12°C