PowerEase™ Touch 600W Power Supply, 115 VAC, 1 each - FAQs
PowerEase™ Touch 600W Power Supply, 115 VAC, 1 each - FAQs
View additional product information for PowerEase™ Touch Power Supply - FAQs (PS0600, PS0120, PS0350)
7 product FAQs found
Can I use the PowerEase Touch 120W Power Supply to transfer two midi gels with the SureLock Tandem Midi Gel Tank and SureLock Tandem Midi Blot Module?
Unfortunately, the PowerEase Touch 120W power supply has a maximum output of 500 mA and is not suitable for transferring two midi gels. For wet transfer, we recommend using the PowerEase Touch Power Supply 350W or 600W unit, which can deliver up to 3 A. More information about these power supply units can be found on our website.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Which applications can be run using the PowerEase Touch 600W Power Supply?
The PowerEase Touch 600W Power Supply is ideal for high power and high current applications such as high-throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, IEF, and western blotting.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How many gels can I simultaneously transfer using the PowerEase Touch 600W Power Supply?
The PowerEase Touch 600W Power Supply can be used for simultaneous transfer of up to 8 mini or 4 midi gels.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
The minimum output current of the PowerEase Touch 120W, 350W, and 600W power supplies is 1 mA, however, I can only set the current to a minimum of 10 mA. Why is this?
The PowerEase Touch 120W, 350W, and 600W power supplies can deliver down to 1 mA. However, because the unit displays power in Amps, with 0.01 A (10 mA) being the minimum display, when the current drops below 5 mA, the power supply will display 0 A but will actually keep running.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How many gels can I simultaneously run using the PowerEase Touch 600W Power Supply?
The PowerEase Touch 600W Power Supply can be used for simultaneous running of up to 16 mini gels or 14 midi gels.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can the PowerEase Touch 600W Power Supply be used to run ZOOM IPG strips?
No, ZOOM IPG strips require a voltage ramp up to 2000 V to be run. We recommend using the PowerEase Touch HV Power Supply to run ZOOM IPG Strips.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I use the PowerEase Touch 120W or PowerEase Touch 350W Power Supply to run isoelectric focusing (IEF) gels?
The voltage required for running of IEF gels cannot be reached with the PowerEase Touch 120W or the PowerEase Touch 350W Power Supply. We recommend using the PowerEase Touch 600W Power Supply or PowerEase Touch HV Power Supply for running IEF gels.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.