Lysis Solution for GeneJET Plasmid Miniprep Kit - FAQs
Lysis Solution for GeneJET Plasmid Miniprep Kit - FAQs
View additional product information for Lysis Solution for GeneJET Plasmid Miniprep Kit - FAQs (R1223)
1 product FAQs found
Do you offer the Lysis Solution from the GeneJET Plasmid Miniprep Kit (Cat. No. K0501, K0503) as a stand-alone product?
Yes, the Lysis Solution from the GeneJET Plasmid Miniprep Kit is available as a stand-alone product (Cat. No. R1223).