A.C.T.™ I Culture Transport
Thermo Scientific™
A.C.T.™ I Culture Transport
Thermo Scientific™ Remel A.C.T. I is an aerobic, facultative and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.
Have Questions?
Número de catálogoCantidad
R12401-MBD20/paq
Número de catálogo R12401-MBD
Precio (MXN)
-
Cantidad:
20/paq
Thermo Scientific™ Remel A.C.T. I is an aerobic, facultative, and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.
  • With semi-solid, nonnutritive medium for use with swab or fluid samples
  • Fluids may be introduced through septum in cap
Especificaciones
DescripciónA.C.T. I
Para utilizar con (aplicación)Healthcare
Cantidad20/paq
Tamaño del tubo16 x 125 mm
Unit SizeEach