Product Image
Thermo Scientific™
Remel™ Sputagest 50
Thermo Scientific™ Remel Sputagest 50 is for digestion and decontamination procedures for AFB and fungal cultures.
Have Questions?
KatalognummerMenge
R210966 x 100 ml
Katalognummer R21096
Preis (EUR)
-
Menge:
6 x 100 ml
Thermo Scientific™ Remel Sputagest 50 is for digestion and decontamination procedures for AFB and fungal cultures.
  • For digestion and decontamination procedures for AFB and fungal cultures
  • For use with 50mL TB Base Digestant or Sodium Citrate
Specifications
BeschreibungSputagest 100 Mucolytic Agent
Menge6 x 100 ml
FormFlüssig
Unit SizeEach