BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid, Aegis™ 5-14 film, 0 ports
Thermo Scientific™
BioProcess Container (BPC), 50L, for HyPerforma™ Single-Use Mixer, liquid-to-liquid, Aegis™ 5-14 film, 0 ports
The 50L Thermo Scientific™ Bioprocess Containers (BPC) for liquid-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic filmWeitere Informationen
Have Questions?
KatalognummerMenge
SH30983.01Jeweils
Katalognummer SH30983.01
Preis (EUR)
-
Menge:
Jeweils
The 50L Thermo Scientific™ Bioprocess Containers (BPC) for liquid-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic film and includes 0 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Anschlussdetail0 ports
ProduktlinieHyPerforma
MengeJeweils
Kapazität (metrisch)50 L
Unit Size50 L