2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors, injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors, injection site

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layerWeitere Informationen
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerKapazität (metrisch)
SS00028I
auch als SS00028-I bezeichnet
2 l
SS00024I
auch als SS00024-I bezeichnet
100 ml
SS00025I
auch als SS00025-I bezeichnet
250 ml
SS00026I
auch als SS00026-I bezeichnet
500 ml
SS00027I
auch als SS00027-I bezeichnet
1 l
Katalognummer SS00028I
auch als SS00028-I bezeichnet
Preis (EUR)
-
Kapazität (metrisch):
2 l

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 2: Luer lock body connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 3: Tubing: EVA, injection port

Specifications
Abmessungen30.5 x 20.3 cm (12.0 x 8.0 in.)
AußenverpackungEin 4 ml-Polybeutel
ProduktlinieLabtainer
Menge50/Cs.
VersandbedingungRaumtemperatur
SterilisationsmethodeBestrahlt
Kapazität (metrisch)2 l
Film DescriptionASI 26/77
MaterialPolyethylen
Port DetailLuer Lock Body Connector and Injection Site
Tubing StyleTPE and EVA
Unit Size2 L
Inhalt und Lagerung
Bei Raumtemperatur lagern.