2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert connectors and injection site
The 10 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, aWeitere Informationen
Have Questions?
Ansicht ändern
Katalognummer
Kapazität (metrisch)
SS00071I
auch als SS00071-I bezeichnet
5 l
SS00072I
auch als SS00072-I bezeichnet
10 l
SS00073I
auch als SS00073-I bezeichnet
20 l
SS00074I
auch als SS00074-I bezeichnet
50 l
4 Optionen
Katalognummer SS00071I
auch als SS00071-I bezeichnet
Preis (EUR)
-
Kapazität (metrisch):
5 l
The 10 L Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (2 MPC insert + cap, 1 break-off injection site).
Features of the system include: • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications include: • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters • Preparing and storing buffer, culture media, and process liquid • Collecting chromatography feed and fraction • Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology Choose from the industry’s largest component library, with over 2,000 unique components that include: • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors • Filters (used for bioburden reduction)—sterilization grade filters and vent filters • Tubing—thermoplastic elastomers and platinum-cured silicone • Sample ports—septum-style and needle-free ports • Clamps—sliders, pinch, and crimp bands
Specifications
Abmessungen31.5 cm x 32.0 cm (12.4 in. x 12.6 in.)
AußenverpackungEin 4 ml-Polybeutel
ProduktlinieLabtainer
Menge30 per case
RichtliniencGMP
VersandbedingungRaumtemperatur
SterilisationsmethodeBestrahlt
Kapazität (metrisch)5 l
Film DescriptionASI 28
MaterialEVA
Port DetailMPC insert connectors and injection site