Product Image
HyPerforma™ Single-Use Mixer (S.U.M.), 100L, Jacketed, 240 VAC, AC Motor, with Load Cell
Have Questions?
Número do catálogo SUM01009006
conhecido também como SUM0100.9006
Preço (BRL)
-
The 100 L Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.), jacketed, 240VAC, AC motor, with load cell, is designed using proven engineered mixer platform principles and is used for upstream and downstream mixing applications. The HyPerforma S.U.M. offers a more ergonomic and efficient user experience. The complete line of HyPerforma S.U.M.s includes 50, 100, 200, 500, 1,000 and 2,000 L sizes that helps ensure consistent scalability while providing a number of application usages from powder media and buffer hydration to advanced downstream harvest and intermediate pooling steps.
Features:
• Unique proprietary mixing system that provides conventional overhead mixing while maintaining sterility and integrity of the S.U.B. BioProcess Container (BPC).
• Minimized vessel footprint and reduced hold-up volume.
• Simplified bag loading with a vertical access door (available on 500 L, 1,000 L sizes and on 2,000 L S.U.M.).
• Cable management system provides greater organization for electrical cables, tubing and line sets.
• Dedicated adjustable tool holder keeps necessary tools available for convenience.
• Mixer vessel is offered with non-jacketed (no heat transfer) or water jacket (vessel side and bottom). Water jacket to be used with separate external temperature control unit for heat transfer requirement.
• Offered with choice of AC motor or DC motor with precise agitation control. Brushless DC motor includes encoder feedback to help improve RPM accuracy and is GFCI-compatible. (Electrical box not available with DC motors).
• Three load cells weighing systems enable accurate batch weight monitoring.
• Option to use probes for temperature, pH, conductivity and pressure monitoring.
• Electrical box with communication cable that provides output signals to customer plant controller.
• S.U.M. BPCs offered in CX5-14 and Aegis5-14 Films.
Applications of the Thermo Scientific HyPerforma S.U.M. include:
• Powder media hydration.
• Liquid media concentrate dilution.
• Harvest hold tank.
• Viral inactivation.
• Downstream intermediate pooling.
• Final product hold and additive suspension.
Related products:
Hyperforma™ Single-Use Mixers (50 L)
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Non-Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (100 L)
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Non-Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Non-Jacketed, 240 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Jacketed, 240 VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (200 L)
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (500 L)
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (1,000 L)
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (2,000 L)
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Jacketed, 240VAC, AC Motor, without Load Cell
Especificações
Unit Size100 L