Product Image
HyPerforma™ Single-Use Mixer (S.U.M.), 200L, Non-Jacketed, DC Motor, without Load Cell
Have Questions?
Catalog number SUM02009001
also known as SUM0200.9001
Price (USD)
-
The 200 L Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.), non-jacketed, DC motor, w/o load cell, is designed using proven engineered mixer platform principles and is used for upstream and downstream mixing applications. The HyPerforma S.U.M. offers a more ergonomic and efficient user experience. The complete line of HyPerforma S.U.M.s includes 50, 100, 200, 500, 1,000 and 2,000 L sizes that helps ensure consistent scalability while providing a number of application usages from powder media and buffer hydration to advanced downstream harvest and intermediate pooling steps.
Features:
• Unique proprietary mixing system that provides conventional overhead mixing while maintaining sterility and integrity of the S.U.B. BioProcess Container (BPC).
• Minimized vessel footprint and reduced hold-up volume.
• Simplified bag loading with a vertical access door (available on 500 L, 1,000 L sizes and on 2,000 L S.U.M.).
• Cable management system provides greater organization for electrical cables, tubing and line sets.
• Dedicated adjustable tool holder keeps necessary tools available for convenience.
• Mixer vessel is offered with non-jacketed (no heat transfer) or water jacket (vessel side and bottom). Water jacket to be used with separate external temperature control unit for heat transfer requirement.
• Offered with choice of AC motor or DC motor with precise agitation control. Brushless DC motor includes encoder feedback to help improve RPM accuracy and is GFCI-compatible. (Electrical box not available with DC motors).
• Three load cells weighing systems enable accurate batch weight monitoring.
• Option to use probes for temperature, pH, conductivity and pressure monitoring.
• Electrical box with communication cable that provides output signals to customer plant controller.
• S.U.M. BPCs offered in CX5-14 and Aegis5-14 Films.
Applications of the Thermo Scientific HyPerforma S.U.M. include:
• Powder media hydration.
• Liquid media concentrate dilution.
• Harvest hold tank.
• Viral inactivation.
• Downstream intermediate pooling.
• Final product hold and additive suspension.
Related products:
Hyperforma™ Single-Use Mixers (50 L)
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Non-Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 50 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (100 L)
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Non-Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Jacketed, 120 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Non-Jacketed, 240 VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 100 L, Jacketed, 240 VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (200 L)
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 200 L, Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (500 L)
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 500 L, Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (1,000 L)
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Non-Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Jacketed, DC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 1000 L, Jacketed, 240VAC, AC Motor, without Load Cell
Hyperforma™ Single-Use Mixers (2,000 L)
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Non-Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Jacketed, 120VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Non-Jacketed, 240VAC, AC Motor, without Load Cell
HyPerforma Single-Use Mixer (S.U.M.), 2000 L, Jacketed, 240VAC, AC Motor, without Load Cell
Specifications
Unit Size200 L