FM&trade; 5-95 (<i>N</i>-(3-Trimethylammoniumpropyl)-4-(6-(4-(Diethylamino)phenyl)hexatrienyl)Pyridinium Dibromide)
Invitrogen™
FM™ 5-95 (N-(3-Trimethylammoniumpropyl)-4-(6-(4-(Diethylamino)phenyl)hexatrienyl)Pyridinium Dibromide)
Die zunehmende Anzahl erfolgreicher Anwendungen für unsere FM-Farbstoffe hat uns dazu veranlasst, FM 5-95 zu synthetisieren, ein etwas weniger lipophilesWeitere Informationen
Have Questions?
KatalognummerMenge
T233601 mg
Katalognummer T23360
Preis (EUR)
876,00
1 mg
Menge:
1 mg
Preis (EUR)
876,00
1 mg
Die zunehmende Anzahl erfolgreicher Anwendungen für unsere FM-Farbstoffe hat uns dazu veranlasst, FM 5-95 zu synthetisieren, ein etwas weniger lipophiles Analogon von FM 4-64 (T-3166, T-13320) mit praktisch identischen spektroskopischen Eigenschaften.
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
FarbeRot
NachweisverfahrenFluoreszent
Zur Verwendung mit (Geräte)Fluoreszenzmikroskop
ProduktlinieFM
Menge1 mg
VersandbedingungRaumtemperatur
MarkertypFluorescent Dye
ProdukttypFarbstoff
Sub Cellular LocalizationPlasmamembran
Unit Size1 mg
Inhalt und Lagerung
Lagerung bei Raumtemperatur und vor Licht geschützt
Häufig gestellte Fragen (FAQ)
I want to study endosome trafficking using FM 4-64 dye. Will the label be retained after fixation? And can I label cells that have already been fixed?
No. For that, you would need the FM 4-64FX version. The non-FX version will be lost, leading to loss of much of the specific label and a vast increase in background labeling. The FX version will be fixed in place with formaldehyde. Cells that have been fixed already will not label correctly, so you will need to label the cells live and then fix.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Zitierungen und Referenzen (14)
Zitierungen und Referenzen
Abstract
Glutamate receptor dynamics organizing synapse formation in vivo.
Authors:Rasse TM, Fouquet W, Schmid A, Kittel RJ, Mertel S, Sigrist CB, Schmidt M, Guzman A, Merino C, Qin G, Quentin C, Madeo FF, Heckmann M, Sigrist SJ
Journal:Nat Neurosci
PubMed ID:16136672
'Insight into how glutamatergic synapses form in vivo is important for understanding developmental and experience-triggered changes of excitatory circuits. Here, we imaged postsynaptic densities (PSDs) expressing a functional, GFP-tagged glutamate receptor subunit (GluR-IIA(GFP)) at neuromuscular junctions of Drosophila melanogaster larvae for several days in vivo. New PSDs, associated with functional ... More
Fluorescence ratio imaging microscopy shows decreased access of vancomycin to cell wall synthetic sites in vancomycin-resistant Staphylococcus aureus.
Authors:Pereira PM, Filipe SR, Tomasz A, Pinho MG,
Journal:Antimicrob Agents Chemother
PubMed ID:17646417
'A new method of fluorescence ratio imaging microscopy was used to compare the in vivo binding capacity and the access of a fluorescent derivative of vancomycin to the cell wall synthetic sites in isogenic pairs of vancomycin-susceptible and -resistant laboratory mutants and vancomycin-intermediate and -susceptible clinical isolates of Staphylococcus aureus. ... More
Single synaptic vesicle tracking in individual hippocampal boutons at rest and during synaptic activity.
Authors:Lemke EA, Klingauf J
Journal:J Neurosci
PubMed ID:16306416
'How synaptic vesicles move within central nervous synapses to their docking sites at the plasma membrane is widely discussed in synaptic physiology. This question is especially difficult to investigate in the small hippocampal boutons, which themselves can slowly move during observation in primary cell culture. Here, we describe a single ... More
SynCAM, a synaptic adhesion molecule that drives synapse assembly.
Authors:Biederer T, Sara Y, Mozhayeva M, Atasoy D, Liu X, Kavalali ET, Südhof TC
Journal:Science
PubMed ID:12202822
'Synapses, the junctions between nerve cells through which they communicate, are formed by the coordinated assembly and tight attachment of pre- and postsynaptic specializations. We now show that SynCAM is a brain-specific, immunoglobulin domain-containing protein that binds to intracellular PDZ-domain proteins and functions as a homophilic cell adhesion molecule at ... More
Lipid spirals in Bacillus subtilis and their role in cell division.
Authors:Barák I, Muchová K, Wilkinson AJ, O'Toole PJ, Pavlendová N,
Journal:Mol Microbiol
PubMed ID:18430139
The fluid mosaic model of membrane structure has been revised in recent years as it has become evident that domains of different lipid composition are present in eukaryotic and prokaryotic cells. Using membrane binding fluorescent dyes, we demonstrate the presence of lipid spirals extending along the long axis of cells ... More