TSE Series -86°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capcity, holds 320 boxes, 120V/60Hz, 16A, painted galvaneel interior - FAQs
TSE Series -86°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capcity, holds 320 boxes, 120V/60Hz, 16A, painted galvaneel interior - FAQs
View additional product information for TSE Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (TSE320A)