TSG Series Glass Door Laboratory Refrigerators
Thermo Scientific™

TSG Series Glass Door Laboratory Refrigerators

The TSG Glass Door Refrigerators offer sustainability at 2-8°C, an outstanding 4-year parts and labor warranty, and tight temperature uniformity in single and double door configurations. All TSG Series utilize SNAP and F-Gas compliant refrigeration and are made at a zero waste to landfill facility.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerKapazität (angloamerikanisch)InnenElektrische Anforderungen
TSG4505GV45,8 cu. ft.Weißer Lack230 V/50 Hz
TSG1205GA12 ft.3Weißer Lack115 V/60 Hz
TSG1205GV12 ft.3Weißer Lack230 V/50 Hz
TSG1205GD12 ft.3Weißer Lack208 bis 230 V/60 Hz
TSG2305GA23 cu. ft.Weißer Lack115 V/60 Hz
TSG2305GV23 cu. ft.Weißer Lack230 V/50 Hz
TSG2305GD23 cu. ft.Weißer Lack208 bis 230 V/60 Hz
TSG3005GA29,2 cu. ft.Weißer Lack115 V/60 Hz
TSG3005GV29,2 cu. ft.Weißer Lack230 V/50 Hz
TSG3005GD29,2 cu. ft.Weißer Lack208/230 V, 50 Hz
TSG4505GA45,8 cu. ft.Weißer Lack115 V/60 Hz
TSG4505GD45,8 cu. ft.Weißer Lack208 bis 230 V/60 Hz
TSG5005GA51,1 cu. ft.Weißer Lack115 V/60 Hz
TSG5005GV51,1 cu. ft.Weißer Lack230 V/50 Hz
TSG5005GD51,1 cu. ft.Weißer Lack208 bis 230 V/60 Hz
Katalognummer TSG4505GV
Preis (EUR)
-
Kapazität (angloamerikanisch):
45,8 cu. ft.
Innen:
Weißer Lack
Elektrische Anforderungen:
230 V/50 Hz
Thermo Scientific TSG Series Glass Door Laboratory Refrigerators offer quiet, intuitive operation with excellent temperature performance and an outstanding warranty, meeting the needs of a wide variety of professional users who need consistent, reliable performance without sacrificing usability or sustainability. All TSG Series Laboratory Refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigerants and are configured to provide excellent temperature performance while minimizing power consumption, all delivered from our zero waste to landfill facility in Asheville, North Carolina.

The drive for sample integrity and security

  • Designed to safely store media, reagents, and other laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested and certified to conform with the design and safety standards of UL 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned glass doors with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Extra and Alternate wire baskets, shelves, and drawers
  • Stainless Steel interior finish available
  • E-Lock Adapter kits
Specifications
ZugriffssicherheitTürschloss
SchrankmaterialPainted Steel
Fassungsvermögen (angloamerikanisch) Einschub100 lb.
Fassungsvermögen (metrisch) Einschub45 kg
RegelungsartCapacitive Touch
BeschreibungTSG Series General Purpose Refrigerator
Abmessungen (T x B x H) außen36.6 x 56.5 x 78.5 in. (931 x 1435 x 1994 mm)
Abmessungen (T x B x H) innen26.0 x 52.5 x 58.0 in. (660 x 1334 x 1473 mm)
Elektrische Anforderungen230 V/50 Hz
Oberflächenbehandlung außenLackiert
Frequenz50 Hz
InnenWeißer Lack
ÜberwachungsoptionenSmartVue Pro, Remote Alarm Contacts
Anschlüsse1 Rear Access Port
KältemittelR290
Einschübe8 Einstellbar
Versandgewicht (angloamerikanisch)611 lb.
Versandgewicht (metrisch)277 kg
Temperatur (Sollwert)+5°C
TypGeneral Purpose Refrigerator
Gewährleistung4 Years Parts and Labor + 1 Year Compressor
Kapazität (angloamerikanisch)45,8 cu. ft.
Kapazität (metrisch)1297 L
Zertifizierung/KonformitätCE
KompressorHermetic
Ausführung der TürSliding Glass
No. of Doors2
SteckertypCEE 7/7
ModellUpright
Temperaturbereich3°C bis 7°C
Spannung208/230 V
Unit SizeEach