Thermo Scientific™

Sample Racks for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors

Catalog number: 6820.4087
Thermo Scientific™

Sample Racks for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors

Catalog number: 6820.4087

Support exceptional sample format versatility with Thermo Scientific™ Dionex™ Sample Racks used for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors, for the WPS-3000 series UHPLC and LC samplers.

 
Catalog Number
6820.4087
Unit Size
Each
Description
Sample rack for 40 vials, 1.1 mL, conical
Price (ARS)
Full specifications
DescriptionSample rack for 40 vials, 1.1 mL, conical
For Use With (Equipment)1.1 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.1 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 40 vials, 1.8 mL, cylindrical
Price (ARS)
Full specifications
DescriptionSample rack for 40 vials, 1.8 mL, cylindrical
For Use With (Equipment)1.8 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.8 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 10 vials, 10 mL, cylindrical
Price (ARS)
Full specifications
DescriptionSample rack for 10 vials, 10 mL, cylindrical
For Use With (Equipment)10 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity10 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 22 vials, 4 mL, cylindrical
Price (ARS)
Full specifications
DescriptionSample rack for 22 vials, 4 mL, cylindrical
For Use With (Equipment)4 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity4 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 72 vials, 1.2 mL, cylindrical
Price (ARS)
Full specifications
DescriptionSample rack for 72 vials, 1.2 mL, cylindrical
For Use With (Equipment)1.2 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.2 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 72 vials, 0.3 mL, cylindrical
Price (ARS)
Full specifications
DescriptionSample rack for 72 vials, 0.3 mL, cylindrical
For Use With (Equipment)0.3 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity0.3 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 40 Eppendorf tubes, 1.5 mL
Price (ARS)
Full specifications
DescriptionSample rack for 40 Eppendorf tubes, 1.5 mL
For Use With (Equipment)1.5 mL Eppendorf Tubes
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.5 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 40 Eppendorf tubes, 0.5 mL
Price (ARS)
Full specifications
DescriptionSample rack for 40 Eppendorf tubes, 0.5 mL
For Use With (Equipment)0.5 mL Eppendorf Tubes
Product LineDionex
TypeAutosampler Rack
Sample Capacity0.5 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Sample rack for 72 micro-dialyse vials, 0.3 mL, cylindrical
Price (ARS)
Full specifications
DescriptionSample rack for 72 micro-dialyse vials, 0.3 mL, cylindrical
For Use With (Equipment)0.3 mL Micro-Dialyse Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity0.3 mL
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDescriptionPrice (ARS)
6820.4087Full specifications
EachSample rack for 40 vials, 1.1 mL, conicalContact Us ›
DescriptionSample rack for 40 vials, 1.1 mL, conical
For Use With (Equipment)1.1 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.1 mL
Unit SizeEach
6820.4070Full specifications
EachSample rack for 40 vials, 1.8 mL, cylindricalContact Us ›
DescriptionSample rack for 40 vials, 1.8 mL, cylindrical
For Use With (Equipment)1.8 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.8 mL
Unit SizeEach
6820.4086Full specifications
EachSample rack for 10 vials, 10 mL, cylindricalContact Us ›
DescriptionSample rack for 10 vials, 10 mL, cylindrical
For Use With (Equipment)10 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity10 mL
Unit SizeEach
6820.4084Full specifications
EachSample rack for 22 vials, 4 mL, cylindricalContact Us ›
DescriptionSample rack for 22 vials, 4 mL, cylindrical
For Use With (Equipment)4 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity4 mL
Unit SizeEach
6820.4090Full specifications
EachSample rack for 72 vials, 1.2 mL, cylindricalContact Us ›
DescriptionSample rack for 72 vials, 1.2 mL, cylindrical
For Use With (Equipment)1.2 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.2 mL
Unit SizeEach
6820.4091Full specifications
EachSample rack for 72 vials, 0.3 mL, cylindricalContact Us ›
DescriptionSample rack for 72 vials, 0.3 mL, cylindrical
For Use With (Equipment)0.3 mL Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity0.3 mL
Unit SizeEach
6820.4094Full specifications
EachSample rack for 40 Eppendorf tubes, 1.5 mLContact Us ›
DescriptionSample rack for 40 Eppendorf tubes, 1.5 mL
For Use With (Equipment)1.5 mL Eppendorf Tubes
Product LineDionex
TypeAutosampler Rack
Sample Capacity1.5 mL
Unit SizeEach
6820.4096Full specifications
EachSample rack for 40 Eppendorf tubes, 0.5 mLContact Us ›
DescriptionSample rack for 40 Eppendorf tubes, 0.5 mL
For Use With (Equipment)0.5 mL Eppendorf Tubes
Product LineDionex
TypeAutosampler Rack
Sample Capacity0.5 mL
Unit SizeEach
6820.4097Full specifications
EachSample rack for 72 micro-dialyse vials, 0.3 mL, cylindricalContact Us ›
DescriptionSample rack for 72 micro-dialyse vials, 0.3 mL, cylindrical
For Use With (Equipment)0.3 mL Micro-Dialyse Vials
Product LineDionex
TypeAutosampler Rack
Sample Capacity0.3 mL
Unit SizeEach
Showing 9 of 9

Support exceptional sample format versatility with Thermo Scientific™ Dionex™ Sample Racks used for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors, for the WPS-3000 series UHPLC and LC samplers.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text