SalivaReady™ Solution
Applied Biosystems™
SalivaReady™ Solution
SalivaReady Solution is a robust buffer that prepares saliva samples for direct use in RT-qPCR without the need for nucleicMás información
Have Questions?
Número de catálogo A50608
Precio (USD)
-
SalivaReady Solution is a robust buffer that prepares saliva samples for direct use in RT-qPCR without the need for nucleic acid extraction or purification.
• Prepare saliva samples for direct use in RT-qPCR with a single reagent
• Reduce hands-on time and time-to-results compared to standard sample prep workflows
• Increase your sensitivity over other direct-to-PCR workflows
• Enhance your ability to handle sample variability due to matrix effects
Single reagent setup
SalivaReady Solution, when combined in a simple 1:1 ratio with saliva sample, is capable of directly proceeding to RT-qPCR analysis after a short heating protocol.
Short, simple protocol
Saliva samples in SalivaReady Solution require a short heating protocol consisting of 5 min at 62°C, 5 min at 92°C, and 2 min at 4°C. This simplified protocol can be easily accomplished on any 96-well-format thermal cycler. Additionally, SalivaReady Solution is compatible with saliva samples that have been heat-treated in their collection tubes, an optional pre-treatment step to inactivate potentially infectious agents.
Advanced formulation
SalivaReady Solution combines the benefits of physical, chemical, and enzymatic treatment solutions in a single formulation to improve RT-qPCR sensitivity and reduce sample variability typically caused by matrix effects and inhibitors.
Ready for high throughput
Up to 96 samples can be prepared at once in a single 96-well plate. Combined with an easy workflow consisting of two easy transfer steps and a short temperature protocol, SalivaReady Solution is ready for high throughput configurations.
Recommended RT-qPCR Master Mixes
TaqPath 1-Step RT-qPCR Master Mix
TaqPath 1-Step Multiplex Master Mix no ROX
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
CompatibilidadRT-qPCR, qPCR
Para utilizar conTaqPath 1-Step RT-qPCR Master Mix (A15299, A15300), TaqPath 1-Step Multiplex Master Mix no ROX (A28521, A28522, A28523)
Para utilizar con (equipo)Thermal Cycler
Compatibilidad de alto rendimientoHigh Throughput-Compatible
N.º de reacciones2000 reacciones
Cantidad2000 reactions
Tipo de muestraSaliva
Condiciones de envíoDry Ice
Volumen de material de partida20 μL
Volumen40 mL
Para utilizar con (aplicación)Real Time PCR (qPCR), RT-PCR
FormularioLíquido
Velocidad de reacciónFast
Unit Size2000 reactions
Contenido y almacenamiento
SalivaReady Solution (40 mL), 2,000 reactions at 20 μL/reaction, store at -20°C