Thermo Scientific™
CETAC™ Autosamplers
Catalog number: 1169950
Thermo Scientific™
CETAC™ Autosamplers
Catalog number: 1169950
Increase the sample throughput of your laboratory with Thermo Scientific™ CETAC™ Autosamplers. Automate the analysis of your solutions with a CETAC ASX-260 or ASX-520 autosampler interfaced with a UV-Vis spectrophotometer. An autosampler configuration provides continuous automated analysis of samples without the need for operator intervention. Thermo Scientific autosamplers feature a metal-free liquid flow path and corrosion resistant coating on all metal parts and can withstand the harshest chemical environments while maintaining unsurpassed accuracy.
Catalog Number
1169950
Unit Size
Each
Description
Cetac ASX-260 Autosampler
Price (BRL)
Full specifications
TypeAutosampler
Hertz50/60 Hz
DescriptionCetac ASX-260 Autosampler
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Cetac ASX-520 Autosampler
Price (BRL)
Full specifications
TypeAutosampler
Hertz50/60 Hz
DescriptionCetac ASX-520 Autosampler
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (BRL)
1169950Full specifications
EachCetac ASX-260 AutosamplerContact Us ›
TypeAutosampler
Hertz50/60 Hz
DescriptionCetac ASX-260 Autosampler
Unit SizeEach
4202467Full specifications
EachCetac ASX-520 AutosamplerContact Us ›
TypeAutosampler
Hertz50/60 Hz
DescriptionCetac ASX-520 Autosampler
Unit SizeEach
Showing 2 of 2
Autosamplers can be interfaced with either Thermo Scientific VISIONlite™ Auto or VISIONlite™ Auto SE software for maximum data security.Adjustable parameters include:
  • Use different rack sizes simultaneously
  • The number of samples measured before the probe is washed
  • Wash and rinse times for the sample probe
  • Shake feature for dislodging droplets from the sample probe
  • The CETAC ASX-260 features a 2-rack configuration for up to 180 samples using 8-ml vials
  • The CETAC ASX-520 features a 4-rack configuration for up to 360 samples using 8-ml vials
Required Accessories: When used with a UV-Visible spectrophotometer and autosampler requires a sipper accessory and autosampler software.