Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (BRL) | Availability | Quantity | |
---|---|---|---|---|---|
19432805L | 5 L | Price: 838.252,80 Your price: | *** | ||
194328005 | 5 mL | Price: 7.327,96 Your price: | *** | ||
194328010 | 10 mL | Price: 13.187,67 Your price: | *** | ||
194328050 | 50 mL | Price: 43.914,39 Your price: | *** | ||
1943280250 | 250 mL | Price: 89.368,01 Your price: | *** | ||
19432801L | 1 L | Price: 167.513,70 Your price: | *** | ||
194328010L | 10 L | Contact Us › | *** | - |
CaptureSelect™ FcXL Affinity Matrix has been specifically designed for the purification of recombinant human IgG, Fc fusion proteins, and plasma derived IgG. It recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4).
Features of this affinity matrix include:
CaptureSelect™ FcXL Affinity Matrix purifies IgGs and Fc fusion proteins directly from complex source materials in a single step with high purity and yield.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main Characteristics: