Vanadium(V)-oxid, 99.6 % min., Thermo Scientific Chemicals
Vanadium(V)-oxid, 99.6 % min., Thermo Scientific Chemicals
Vanadium(V)-oxid, 99.6 % min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(V)-oxid, 99.6 % min., Thermo Scientific Chemicals

CAS: 1314-62-1 | O5V2 | 181.88 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
100 g
500 g
Katalognummer 011094.36
auch als 011094-36 bezeichnet
Preis (EUR)
199,00
Each
Menge:
500 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
199,00
Each
Chemikalien-Kennzeichnungen
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormGranules
Assay from Supplier's CofA≥99.6% (V₂O₅)
Appearance (Color)Yellow-orange
Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst for the manufacture of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms. Additionally, it is used as ceramics colorant, as dye and also as a detector material in bolometers in the field of thermal imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only