Thermo Scientific Chemicals

Silberjodid, Premion™, 99.999 % (Metallbasis), Thermo Scientific Chemicals

Katalognummer: 012111.06
5 g, Each
Thermo Scientific Chemicals

Silberjodid, Premion™, 99.999 % (Metallbasis), Thermo Scientific Chemicals

Katalognummer: 012111.06
5 g, Each
Menge
Katalognummer: 012111.06
auch als 012111-06 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
7783-96-2
IUPAC Name
silver(1+) iodide
Molecular Formula
AgI
InChI Key
MSFPLIAKTHOCQP-UHFFFAOYSA-M
SMILES
[Ag+].[I-]
Total Metal Impurities
0.001% max.

Beschreibung

Silver iodide is used in photographic film. Used in medical devices as a sustained release antibacterial. It also used in as an antiseptic and in cloud seeding. It is used in proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P273 - Avoid release to the environment