Natriumtetraborat, wasserfrei, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Natriumtetraborat, wasserfrei, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Natriumtetraborat, wasserfrei, 99.5 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Natriumtetraborat, wasserfrei, 99.5 % (Metallbasis), Thermo Scientific Chemicals

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 kg
5 kg
Katalognummer 012305.A1
auch als 012305-A1 bezeichnet
Preis (EUR)
63,20
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
63,20
Each
Chemikalien-Kennzeichnungen
CAS1330-43-4
IUPAC Namedisodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular FormulaB4Na2O7
InChI KeyNXSVMNYRVBQCMO-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
FormPowder or crystalline powder or crystals or granular
Total Metal Impurities≤0.5%
Sodium tetraborate is used as a co-catalyst for (2,2,6,6-tetramethylpiperidin-1-yl)oxyl and sodium hypochlorite oxidation of alcohols in greener and non-chlorinated solvents. It acts as a reagent for the decomposition of oxides and silicates by fusion. It is used to prepare buffers, which is used in gel electrophoresis of DNA. It acts as a source of borate, micronutrient fertilizer, preservative in taxidermy, additive in ceramic slips and glazes. It is also used as a precursor for sodium perborate monohydrate and boric acid. It finds application as fluoride detoxification, neutron absorber in nuclear reactors and to make indelible ink for dip pens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only