Strontiumchlorid-Hexahydrat, technischer Gütegrad, Thermo Scientific Chemicals
Strontiumchlorid-Hexahydrat, technischer Gütegrad, Thermo Scientific Chemicals
Strontiumchlorid-Hexahydrat, technischer Gütegrad, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontiumchlorid-Hexahydrat, technischer Gütegrad, Thermo Scientific Chemicals

CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
2 kg
Katalognummer 012494.36
auch als 012494-36 bezeichnet
Preis (EUR)
57,70
Each
Menge:
500 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
57,70
Each
Chemikalien-Kennzeichnungen
CAS10025-70-4
IUPAC Namestrontium(2+) hexahydrate dichloride
Molecular FormulaCl2H12O6Sr
InChI KeyAMGRXJSJSONEEG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities≤5.0%
Appearance (Color)White
Assay from Supplier's CofAStrontium: 30.0-35.0%
FormPowder or crystals or chunks
Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the field of medicine, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only