Silizium(IV)-oxid, 99.5 %, Thermo Scientific Chemicals
Silizium(IV)-oxid, 99.5 %, Thermo Scientific Chemicals
Silizium(IV)-oxid, 99.5 %, Thermo Scientific Chemicals
Silizium(IV)-oxid, 99.5 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silizium(IV)-oxid, 99.5 %, Thermo Scientific Chemicals

CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 kg
10 kg
Katalognummer 013024.A9
auch als 013024-A9 bezeichnet
Preis (EUR)
200,00
Each
Zum Warenkorb hinzufügen
Menge:
10 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
200,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormMicrocrystalline powder
Assay (special)Dry Brightness: 84.0-88.7 (TAPPI)
Assay (special)Oil Absorption: 26.0-31.0%
Assay (special)Yelow (B*): 1.6-3.2 (CIEB*)
Particle Size≥97.0% of particles are ≤10 microns
Mehr anzeigen
In the manufacture of glass, ceramics and abrasives, in water filtration, in cosmetics, pharmaceuticals and rubber, as a thermal insulatorSilica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only