Caesiumhydrogencarbonat, 99.99 % (Metallbasis), Thermo Scientific Chemicals
Caesiumhydrogencarbonat, 99.99 % (Metallbasis), Thermo Scientific Chemicals
Caesiumhydrogencarbonat, 99.99 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Caesiumhydrogencarbonat, 99.99 % (Metallbasis), Thermo Scientific Chemicals

CAS: 29703-01-3 | CHCsO3 | 193.92 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
Katalognummer 014234.14
auch als 014234-14 bezeichnet
Preis (EUR)
92,80
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
92,80
Each
Chemikalien-Kennzeichnungen
CAS29703-01-3
IUPAC Namecaesium(1+) hydrogen carbonate
Molecular FormulaCHCsO3
InChI KeyZMCUDHNSHCRDBT-UHFFFAOYSA-M
SMILES[Cs+].OC([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.01% max.
Cesium hydrogen carbonate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only