Kaliumnitrit, ACS, 96.0 % min., Thermo Scientific Chemicals
Kaliumnitrit, ACS, 96.0 % min., Thermo Scientific Chemicals
Kaliumnitrit, ACS, 96.0 % min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kaliumnitrit, ACS, 96.0 % min., Thermo Scientific Chemicals

CAS: 7758-09-0 | KNO2 | 85.10 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
100 g
500 g
Katalognummer 014245.09
auch als 014245-09 bezeichnet
Preis (EUR)
-
Menge:
10 g
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS7758-09-0
IUPAC Namepotassium nitrite
Molecular FormulaKNO2
InChI KeyBXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES[K+].[O-]N=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)96.0% KNO₂ min
Impurity contentCa0.005%
Impurity contentCl 0.03%
Impurity contentFe 0.001%
Impurity contentMg0.002%
Mehr anzeigen
In analytical chemistry (testing for amino acids, cobalt, iodine, urea).Potassium nitrite is used as a food preservative, paint and coating additives and analytical reagent. It is used as a strong oxidizing agent as well as involved in the preparation of heat transfer salts. In addition, it is employed as corrosion inhibitors and anti-scaling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only