Natrium 1-Pentansulfonat-Monohydrat, HPLC-Gütegrad, Thermo Scientific Chemicals
Natrium 1-Pentansulfonat-Monohydrat, HPLC-Gütegrad, Thermo Scientific Chemicals
Natrium 1-Pentansulfonat-Monohydrat, HPLC-Gütegrad, Thermo Scientific Chemicals
Natrium 1-Pentansulfonat-Monohydrat, HPLC-Gütegrad, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Natrium 1-Pentansulfonat-Monohydrat, HPLC-Gütegrad, Thermo Scientific Chemicals

Ionenpaarreagenz
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer 015714.14
auch als 015714-14 bezeichnet
Preis (EUR)
168,00
Each
Zum Warenkorb hinzufügen
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
168,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS207605-40-1
IUPAC Namepentane-1-sulfonate
Molecular FormulaC5H11O3S
InChI KeyRJQRCOMHVBLQIH-UHFFFAOYSA-M
SMILESCCCCCS([O-])(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
Assay (unspecified)≥98.0%
Optical Absorbance at210nm: <0.200 AU ( 0.25M aq. solution)
Optical Absorbance at240nm: <0.050 AU ( 0.25M aq. solution)
Optical Absorbance at250nm: <0.050 AU ( 0.25M aq. solution)
Mehr anzeigen
Ion pair reagent.Sodium 1-pentanesulfonate monohydrate is used as a surfactant and a reversed-phase ion-pairing agent. It is utilized to study its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy. Further, it is used for HPLC analysis of proteins and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only