Barium-Metaphosphat, 90+ %, Thermo Scientific Chemicals
Barium-Metaphosphat, 90+ %, Thermo Scientific Chemicals
Barium-Metaphosphat, 90+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium-Metaphosphat, 90+ %, Thermo Scientific Chemicals

Have Questions?
KatalognummerMenge
022641.36
auch als 022641-36 bezeichnet
500 g
Katalognummer 022641.36
auch als 022641-36 bezeichnet
Preis (EUR)
239,00
Each
Menge:
500 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
239,00
Each
Chemikalien-Kennzeichnungen
CAS13762-83-9
Molecular FormulaBaO6P2
InChI KeyRYSCSJQMVDRYDV-UHFFFAOYSA-J
SMILES[Ba+4].[O-][P]([O-])=O.[O-][P]([O-])=O
Molecular Weight (g/mol)295.27
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)90.1% min
Barium metaphosphate, used in glasses, porcelains, and enamels, as a flame retardant and in phosphors applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only