Strontiumnitrat, wasserfrei, 98 %, Thermo Scientific Chemicals
Strontiumnitrat, wasserfrei, 98 %, Thermo Scientific Chemicals
Strontiumnitrat, wasserfrei, 98 %, Thermo Scientific Chemicals
Strontiumnitrat, wasserfrei, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontiumnitrat, wasserfrei, 98 %, Thermo Scientific Chemicals

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1 kg
5 x 1 kg
Katalognummer 030534.A1
auch als 030534-A1 bezeichnet
Preis (EUR)
76,00
Each
Zum Warenkorb hinzufügen
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
76,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)97.6% min
In railroad flares, signal lights, marine signalsStrontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. Mixed with glycolic acid, it is used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only