Thermo Scientific Chemicals

Lineare Alkylbenzolsulfonsäure, 97 %, Thermo Scientific Chemicals

Katalognummer: 030574.A1
1 kg, Each
Thermo Scientific Chemicals

Lineare Alkylbenzolsulfonsäure, 97 %, Thermo Scientific Chemicals

Katalognummer: 030574.A1
1 kg, Each
Menge
Katalognummer: 030574.A1
auch als 030574-A1 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
68584-22-5
IUPAC Name
4-(dodecan-3-yl)benzene-1-sulfonic acid
Molecular Formula
C18H30O3S
InChI Key
QJRVOJKLQNSNDB-UHFFFAOYNA-N
SMILES
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(O)(=O)=O
Appearance (Color)
Clear dark yellow to dark orange
Form
Viscous liquid
Assay (unspecified)
≥96.0%

Beschreibung

Dodecyl Benzene Sulfonic Acid can be used in all types of household, industrial and institutional cleaners including laundry, dishwash, carwash, hard surface and general purpose detergents. It can also be utilized as an intermediate in preparation of emulsifiers and wetting and foaming agents. Unneutralized compound is an effective surfactant in acidic products such as liquid bowl, aluminum and dairy cleaners. It is an important raw material and intermediate used in Organic Synthesis, Pharma synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician