1-Decen, 96 %, Restisomere, Thermo Scientific Chemicals
1-Decen, 96 %, Restisomere, Thermo Scientific Chemicals
1-Decen, 96 %, Restisomere, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Decen, 96 %, Restisomere, Thermo Scientific Chemicals

CAS: 872-05-9 | C10H20 | 140.27 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
2 kg
Katalognummer 031213.22
auch als 031213-22 bezeichnet
Preis (EUR)
33,70
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
33,70
Each
Chemikalien-Kennzeichnungen
CAS872-05-9
IUPAC Namedec-1-ene
Molecular FormulaC10H20
InChI KeyAFFLGGQVNFXPEV-UHFFFAOYSA-N
SMILESCCCCCCCCC=C
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Clear colorless
Assay (GC)≥95.0%
FormLiquid
1-Decene is the only isomer of industrial importance. it is used as a monomer in copolymers and is an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics. 1-Decene has been isolated from the leaves and rhizome of the plant Farfugium japonicum and has been detected as the initial product in the microbial degradation of n-decane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only