Thermo Scientific Chemicals

N-Octadecan, 99 %, Thermo Scientific Chemicals

Katalognummer: 031954.36
500 g, Each
Thermo Scientific Chemicals

N-Octadecan, 99 %, Thermo Scientific Chemicals

Katalognummer: 031954.36
500 g, Each
Menge
Katalognummer: 031954.36
auch als 031954-36 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
593-45-3
IUPAC Name
octadecane
Molecular Formula
C18H38
InChI Key
RZJRJXONCZWCBN-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC
Appearance (Color)
White
Form
Low melting solid
Assay from Supplier's CofA
>99%

Beschreibung

n-octadecane is used as solvent, lubricant, transformer oil and anti-corrosion agents. It is also used in paraffin as well as chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Aspiration Toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H304 - May be fatal if swallowed and enters airways

    EU Specific Hazard Statements

    EUH066 - Repeated exposure may cause skin dryness or cracking

    Precautionary Statements

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P331 - Do NOT induce vomiting